siRNA Details
Gene Symbol: Igsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177591.4 | NP_808259.2 | 14 | 2569 | ||
NM_183336.2 | NP_899179.1 | 5 | 823 | |||
GenBank mRNA | AK030701.1 | - | 5 | 825 | ||
AK076427.1 | - | 5 | 816 | |||
AK129126.1 | - | 14 | 2551 | |||
AY227771.1 | - | 14 | 2563 | |||
AY227774.1 | - | 5 | 818 | |||
BC119264.1 | - | 5 | 786 | 8734013 | ||
BC119266.1 | - | 5 | 786 | 8734015 | ||
BC144762.1 | - | 5 | 786 |

More Information
Other Species Targeted

Gene Ontology Categories:
