siRNA Details
Gene Symbol: Fbxw13 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177598.3 | NP_808266.1 | 8 | 1102 | ||
GenBank mRNA | AK054339.1 | - | 8 | 1103 | ||
BC119542.1 | - | 8 | 1089 | 40056515 | ||
BC127237.1 | - | 7 | 925 | 40056507 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
