siRNA Details
Gene Symbol: 4933430I17Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177607.3 | NP_808275.1 | 13 | 1236 | ||
GenBank mRNA | AK077205.1 | - | 13 | 1237 | ||
BC116650.1 | - | 13 | 1206 | 40088336 |
