siRNA Details
Gene Symbol: Unc79 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081017.2 | NP_001074486.2 | 44, 45 | 7787 | ||
GenBank mRNA | AB093296.1 | - | 27, 28 | 4875 | ||
AB257853.1 | - | 44, 45 | 7787 | |||
AK033439.1 | - | 7, 8 | 758 | |||
GQ334471.1 | - | 47, 48 | 7266 |

More Information
Other Species Targeted

Gene Ontology Categories:
