siRNA Details
Gene Symbol: Zmym2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_029498.3 | NP_083774.2 | 2 | 450 | ||
GenBank mRNA | AK135499.1 | - | 3 | 563 | ||
BC046507.1 | - | 3 | 496 | |||
BC141403.1 | - | 2 | 426 | 9056016 | ||
BU055093.1 | - | 3 | 564 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: XM_008770810.
Gene Ontology Categories:
