siRNA Details
Gene Symbol: Ric1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK048394.1 | - | 4 | 921 | ||
BC125448.1 | - | 4 | 870 | |||
BC132346.1 | - | 4 | 870 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
