siRNA Details
Gene Symbol: Dgkd | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177646.3 | NP_808314.2 | 30 | 5349 | ||
GenBank mRNA | AK052312.1 | - | 2171 | |||
AK076439.1 | - | 2029 | ||||
AK172901.1 | - | 26 | 4794 | |||
BC026414.1 | - | 4 | 2100 | |||
BC048957.1 | - | 9 | 2805 | |||
BC057061.1 | - | 7 | 2511 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
