siRNA Details
Gene Symbol: Dmxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081371.2 | NP_001074840.2 | 38, 39 | 8453 | ||
GenBank mRNA | AK050888.1 | - | 2, 3 | 157 | ||
AK166520.1 | - | 3, 4 | 207 | |||
AY590892.1 | - | 38, 39 | 8453 | |||
BC056490.1 | - | 15, 16 | 2617 |

More Information
Other Species Targeted

Gene Ontology Categories:
