siRNA Details
Gene Symbol: Slc22a15 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001039371.2 | NP_001034460.2 | 8 | 1472 | ||
GenBank mRNA | AK029299.1 | - | 1 | 935 | ||
AK029695.1 | - | 5 | 630 | |||
BC094925.1 | - | 8 | 1361 | |||
BC152347.1 | - | 3 | 367 |

More Information
Other Species Targeted

Gene Ontology Categories:
