siRNA Details
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038960.1 | - | 4 | 1999 | ||
AK039299.1 | - | 1712 | ||||
BC145738.1 | - | 4 | 1761 | |||
BC145740.1 | - | 4 | 1763 |

UserName
Please enter the information below and press OK to send your cart to Core Services for purchase.
Gene Symbol: AI854703 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK038960.1 | - | 4 | 1999 | ||
AK039299.1 | - | 1712 | ||||
BC145738.1 | - | 4 | 1761 | |||
BC145740.1 | - | 4 | 1763 |