siRNA Details
Gene Symbol: BC048671 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177738.2 | NP_808406.1 | 3, 4 | 671 | ||
GenBank mRNA | AK077180.1 | - | 3, 4 | 673 | ||
BC048671.1 | - | 2, 3 | 385 | 6774036 |

More Information
Other Species Targeted

Gene Ontology Categories:
