siRNA Details
Gene Symbol: Zfp711 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177747.3 | NP_808415.2 | 2, 3 | 486 | ||
GenBank mRNA | AK038956.1 | - | 2, 3 | 488 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
