siRNA Details
Gene Symbol: Kir3dl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177748.2 | NP_808416.1 | 3 | 173 | ||
GenBank mRNA | AF548540.1 | - | 3 | 173 | ||
AF548541.1 | - | 3 | 173 | |||
AK046171.1 | - | 5 | 372 | |||
AY530784.1 | - | 3 | 173 | |||
BC132013.1 | - | 3 | 222 | |||
BC132015.1 | - | 3 | 222 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
