siRNA Details
Gene Symbol: Kif26b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001161665.1 | NP_001155137.1 | 12 | 4721 | ||
GenBank mRNA | AB355846.1 | - | 12 | 4721 | ||
AK076435.1 | - | 1 | 466 | |||
BC056349.1 | - | 7 | 2804 | 6405419 | ||
BC068141.1 | - | 9 | 3533 |
