siRNA Details
Gene Symbol: Prex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177782.3 | NP_808450.2 | 30 | 3903 | ||
GenBank mRNA | AK090301.1 | - | 4 | 381 | ||
AK173168.1 | - | 30 | 3903 | |||
BC057617.1 | - | 5 | 362 | |||
BC067047.1 | - | 28 | 3806 | |||
BC151074.1 | - | 30 | 3851 | 9087987 | ||
BC157940.1 | - | 30 | 3851 | 9088120 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
