siRNA Details
Gene Symbol: Ppfia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205341.1 | NP_001192270.1 | 2 | 477 | ||
NM_177373.4 | NP_796347.2 | 2 | 477 | |||
GenBank mRNA | AK030621.1 | - | 2 | 448 | ||
AK220491.1 | - | 2 | 434 | |||
BC065133.1 | - | 2 | 448 | 30536236 | ||
EU568870.1 | - | 2 | 448 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
