siRNA Details
Gene Symbol: Dmxl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081371.2 | NP_001074840.2 | 42 | 8821 | ||
GenBank mRNA | AK050888.1 | - | 6 | 525 | ||
AK166520.1 | - | 7 | 575 | |||
AY590892.1 | - | 42 | 8821 | |||
BC056490.1 | - | 19 | 2985 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
