siRNA Details
Gene Symbol: CSTF3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001326.2 | NP_001317.1 | 9, 10 | 837 | ||
GenBank mRNA | AK290836.1 | - | 9, 10 | 837 | ||
AK294204.1 | - | 6, 7 | 430 | |||
AK297371.1 | - | 8, 9 | 719 | |||
BC108319.1 | - | 9, 10 | 811 | 6197136 | ||
U15782.1 | - | 9, 10 | 802 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
