siRNA Details
Gene Symbol: Zfp507 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177739.3 | NP_808407.2 | 7 | 3614 | ||
GenBank mRNA | AK038716.1 | - | 1 | 717 | ||
AK129283.1 | - | 7 | 3582 | |||
AK162510.1 | - | 4 | 1904 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
