siRNA Details
Gene Symbol: Wdr66 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK015152.1 | - | 1 | 185 | ||
AK030281.1 | - | 2, 3 | 2357 | |||
BC138176.1 | - | 2, 3 | 489 | 8861195 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
