siRNA Details
Gene Symbol: Elmod1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK029207.1 | - | 11 | 1066 | ||
BC120566.1 | - | 10 | 901 | |||
BC144961.1 | - | 9, 10 | 897 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
