siRNA Details
Gene Symbol: Prex1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177782.3 | NP_808450.2 | 32 | 4063 | ||
GenBank mRNA | AK090301.1 | - | 6 | 541 | ||
AK173168.1 | - | 32 | 4063 | |||
BC057617.1 | - | 7 | 522 | |||
BC067047.1 | - | 30 | 3966 | |||
BC151074.1 | - | 32 | 4011 | 9087987 | ||
BC157940.1 | - | 32 | 4011 | 9088120 |

More Information
Other Species Targeted

Gene Ontology Categories:
