siRNA Details
Gene Symbol: Fam49a | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001146119.1 | NP_001139591.1 | 12 | 3279 | ||
NM_029758.4 | NP_084034.2 | 13 | 3352 | |||
GenBank mRNA | AK036232.1 | - | 1 | 1345 | ||
BC052465.1 | - | 12 | 3260 | 6834768 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
