siRNA Details
Gene Symbol: Pxdn | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_181395.2 | NP_852060.2 | 8 | 856 | ||
GenBank mRNA | AK054255.1 | - | 7 | 771 | ||
AK122223.1 | - | 8 | 626 | |||
AK142872.1 | - | 8 | 856 | |||
AK163597.1 | - | 8 | 843 | |||
BC150789.1 | - | 8 | 851 | 9087700 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
