siRNA Details
Gene Symbol: Zfp429 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001080941.1 | NP_001074410.1 | 3, 4 | 303 | ||
GenBank mRNA | AK083314.1 | - | 4, 5 | 371 | ||
BC046484.1 | - | 3, 4 | 287 |
Gene Symbol: Zfp459 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177811.4 | NP_808479.1 | 3, 4 | 308 | ||
GenBank mRNA | AK036923.1 | - | 3, 4 | 309 | ||
BC100389.1 | - | 3, 4 | 274 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
