siRNA Details
Gene Symbol: Rft1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177815.3 | NP_808483.2 | 4, 5 | 502 | ||
GenBank mRNA | AK086390.1 | - | 4, 5 | 491 | ||
BC082539.1 | - | 4, 5 | 470 | |||
BC094336.1 | - | 4, 5 | 470 | |||
BC116368.2 | - | 4, 5 | 483 | |||
BC116369.2 | - | 4, 5 | 483 | |||
BC141275.1 | - | 4, 5 | 496 | 9055887 | ||
BC145606.1 | - | 4, 5 | 496 | 9054172 |
