siRNA Details
Gene Symbol: Ssh2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001291190.1 | NP_001278119.1 | 14 | 5071 | ||
NM_177710.4 | NP_808378.2 | 15 | 4935 | |||
GenBank mRNA | AK031349.1 | - | 1 | 700 | ||
AK173243.1 | - | 4 | 3799 | |||
BC030940.1 | - | 1 | 1483 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
