siRNA Details
Gene Symbol: Kir3dl2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177748.2 | NP_808416.1 | 9 | 1376 | ||
GenBank mRNA | AF548540.1 | - | 9 | 1376 | ||
AF548541.1 | - | 9 | 1376 | |||
AK046171.1 | - | 11 | 1576 | |||
AY530784.1 | - | 9 | 1376 |
Gene Symbol: Kir3dl1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177749.3 | NP_808417.2 | 9 | 1393 | ||
GenBank mRNA | AY130461.1 | - | 9 | 1380 | ||
AY530782.1 | - | 9 | 1366 | |||
AY530783.1 | - | 9 | 1367 | |||
BB314335.2 | - | 436 | ||||
BC141012.1 | - | 9 | 1379 | 9055624 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
