siRNA Details
Gene Symbol: Gna11 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_010301.3 | NP_034431.1 | 7 | 3212 | ||
GenBank mRNA | AK088773.1 | - | 1 | 1840 | ||
AK143685.1 | - | 7 | 3213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
