siRNA Details
Gene Symbol: Ppfia2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001205341.1 | NP_001192270.1 | 6 | 879 | ||
NM_177373.4 | NP_796347.2 | 6 | 879 | |||
GenBank mRNA | AK030621.1 | - | 6 | 850 | ||
AK220491.1 | - | 6 | 836 | |||
BC065133.1 | - | 6 | 850 | 30536236 | ||
EU568870.1 | - | 6 | 850 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Rat: NM_001108745, XM_008765336, XM_008765332, XM_008765331, XM_008765333, and others.
Gene Ontology Categories:
