siRNA Details
Gene Symbol: Fam179b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177805.3 | NP_808473.2 | 1 | 1326 | ||
GenBank mRNA | AK039995.1 | - | 1 | 1014 | ||
AK141238.1 | - | 1 | 1266 | |||
AK172948.1 | - | 1 | 1254 | |||
BC157958.1 | - | 1 | 1128 | 9088131 | ||
BC158008.1 | - | 1 | 1128 | 9007316 | ||
BC172117.1 | - | 1 | 1128 | 9054403 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
