siRNA Details
Gene Symbol: Sh2d4b | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177816.3 | NP_808484.2 | 6 | 948 | ||
GenBank mRNA | AK050671.1 | - | 3 | 662 | ||
AK134033.1 | - | 3 | 438 | |||
AX776199.1 | - | 6 | 948 | |||
BC138964.1 | - | 6 | 1046 | 8861986 |
