siRNA Details
Gene Symbol: Thada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183021.3 | NP_898842.2 | 3 | 286 | ||
GenBank mRNA | AK044550.1 | - | 3 | 239 | ||
AK154078.1 | - | 3 | 284 | |||
EF222207.1 | - | 3 | 194 |

More Information
Other Species Targeted

Gene Ontology Categories:
