siRNA Details
Gene Symbol: Spink10 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177829.3 | NP_808497.1 | 5, 6 | 457 | ||
GenBank mRNA | AK039062.1 | - | 5, 6 | 458 | ||
AK162840.1 | - | 5, 6 | 425 | |||
BC125360.1 | - | 5, 6 | 373 | 40129875 | ||
BC125362.1 | - | 5, 6 | 373 | 40129877 | ||
DQ437330.1 | - | 5, 6 | 457 |
