siRNA Details
Gene Symbol: Zfp236 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177832.3 | NP_808500.2 | 26 | 4744 | ||
GenBank mRNA | AK041130.1 | - | 4 | 377 | ||
AK082879.1 | - | 3 | 392 | |||
AK156842.1 | - | 6 | 847 | |||
BC036962.1 | - | 2 | 166 | |||
BC138187.1 | - | 26 | 4744 | 8861206 | ||
BC145179.1 | - | 27 | 4888 | |||
BC145180.1 | - | 28 | 4999 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
