siRNA Details
Gene Symbol: Gm15850 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK078387.1 | - | 142 | |||
BC107309.1 | - | 1 | 129 | |||
BC107310.1 | - | 1 | 129 | |||
BC114580.1 | - | 1 | 129 | |||
BC115961.1 | - | 1 | 129 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
