siRNA Details
Gene Symbol: Lcn6 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001276448.1 | NP_001263377.1 | 4 | 428 | ||
NM_177840.4 | NP_808508.2 | 4 | 428 | |||
GenBank mRNA | AK033743.1 | - | 2 | 956 | ||
FJ465143.1 | - | 4 | 405 | |||
FJ465144.1 | - | 4 | 405 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
