siRNA Details
Gene Symbol: Skint9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177864.2 | NP_808532.1 | 3 | 189 | ||
GenBank mRNA | AK037244.1 | - | 3 | 189 | ||
EF494907.1 | - | 3 | 181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
