siRNA Details
Gene Symbol: Skint2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001285963.1 | NP_001272892.1 | 4, 5 | 736 | ||
NM_001285965.1 | NP_001272894.1 | 4, 5 | 736 | |||
GenBank mRNA | AK040156.1 | - | 3, 4 | 725 | ||
EF494892.1 | - | 4, 5 | 736 | |||
EF494893.1 | - | 3, 4 | 736 | |||
EU099299.1 | - | 4, 5 | 736 | |||
EU099300.1 | - | 4, 5 | 736 | |||
EU650234.1 | - | 3, 4 | 710 |

More Information
Other Species Targeted

