siRNA Details
Gene Symbol: Fhad1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177868.4 | NP_808536.2 | 4, 5 | 673 | ||
GenBank mRNA | AK045799.1 | - | 6, 7 | 1003 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
