siRNA Details
Gene Symbol: Adamts3 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001081401.2 | NP_001074870.1 | 3 | 612 | ||
NM_177872.2 | NP_808540.2 | 3 | 612 | |||
GenBank mRNA | AK031900.1 | - | 3 | 613 | ||
BC158091.1 | - | 3 | 213 | 9088176 | ||
BC158105.1 | - | 3 | 213 | 9088181 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
