siRNA Details
Gene Symbol: Zfp82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001253385.1 | NP_001240314.1 | 7 | 826 | ||
GenBank mRNA | AK037211.1 | - | 7 | 823 | ||
AK129467.1 | - | 1 | 983 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
