siRNA Details
Gene Symbol: Gm4884 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183166.2 | NP_898989.2 | 2 | 260 | ||
GenBank mRNA | AK029779.1 | - | 2 | 260 |
Gene Symbol: Gm5114 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177890.3 | NP_808558.2 | 2 | 249 | ||
GenBank mRNA | AK029498.1 | - | 2 | 250 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
