siRNA Details
Gene Symbol: Fan1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177893.3 | NP_808561.2 | 2 | 578 | ||
GenBank mRNA | AK077918.1 | - | 2 | 569 | ||
AK173087.1 | - | 1 | 36 | |||
BC116270.2 | - | 1 | 704 | |||
BC116271.2 | - | 1 | 704 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
