siRNA Details
Gene Symbol: Gm16938 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | AK085722.1 | - | 359 | |||
BC144932.1 | - | 3 | 450 | |||
BC144933.1 | - | 3 | 423 | |||
BC144936.1 | - | 3 | 423 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
