siRNA Details
Gene Symbol: Slc9a9 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177909.5 | NP_808577.3 | 3 | 617 | ||
GenBank mRNA | AK037058.1 | - | 3 | 601 | ||
AK156494.1 | - | 3 | 578 | |||
AK172687.1 | - | 3 | 589 | |||
BC153825.1 | - | 3 | 595 | 40093280 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
