siRNA Details
Gene Symbol: Igsf1 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_177591.4 | NP_808259.2 | 4 | 376 | ||
NM_177915.4 | NP_808583.1 | 4 | 376 | |||
NM_183335.2 | NP_899178.2 | 3 | 240 | |||
GenBank mRNA | AK030452.1 | - | 4 | 348 | ||
AK129126.1 | - | 4 | 358 | |||
AY227771.1 | - | 4 | 370 | |||
AY227772.1 | - | 4 | 370 | |||
AY227773.1 | - | 3 | 240 | |||
BC089474.1 | - | 4 | 357 | |||
BC100420.1 | - | 4 | 376 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
