siRNA Details
Gene Symbol: Bptf | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_176850.2 | NP_789820.2 | 28 | 9306 | ||
GenBank mRNA | AK078162.1 | - | 1 | 325 | ||
AK082852.1 | - | 4 | 877 |

More Information
Other Species Targeted

Gene Ontology Categories:
