siRNA Details
Gene Symbol: 9830107B12Rik | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001177896.1 | NP_001171367.1 | 2 | 512 | ||
NM_177824.4 | NP_808492.1 | 2 | 512 | |||
GenBank mRNA | AK079398.1 | - | 2 | 473 | ||
EF646801.1 | - | 213 |

More Information
Additional Information:
This siRNA no longer meets our design criteria. We suggest you choose a different siRNA for this gene or contact Tech Support for more information.
Gene Ontology Categories:
