siRNA Details
Gene Symbol: Thada | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_183021.3 | NP_898842.2 | 4 | 384 | ||
GenBank mRNA | AK044550.1 | - | 4 | 337 | ||
AK154078.1 | - | 4 | 382 | |||
EF222207.1 | - | 4 | 292 |

More Information
Other Species Targeted

Gene Ontology Categories:
